Learn More
Invitrogen™ Human IgG4 ELISA Kit
Description
The Human ImmunoGlobulin 4 (Hu IgG4) ELISA quantitates Hu IgG4 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG4. Principle of the method The Human IgG4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P01861 |
| Assay Range | 9.4-600 ng/mL |
| Assay Sensitivity | 0.1 ng/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human total IgG HRP-Conjugate anti-human IgG4 monoclonal antibody human total IgG Standard lyophilized, 1,200 ng/mL upon reconstitution Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Blue-Dye Green-Dye Adhesive Films |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.