Learn More
Cayman Chemical IgG4humn Monclonal ntIb 100ug
SDP
Catalog No. 502946169
Encompass_Preferred
Immunogen: Peptide corresponding to the hinge region of human IgG4 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgG4; (-) Human IgG1, IgG2, IgG3, IgM, IgA, IgD, IgE; (-) Cynomolgus monkey, goat, mouse, rat, or rhesus monkey IgG • Applications: ELISA, FC, ICC, IHC, WB • MW = 150 and 50 kDa for whole IgG4 and the IgG4 heavy chain under non-reducing and reducing conditions, respectively
Catalog No. 50-294-6169 Supplier Cayman Chemical Supplier No. 32123100 UG
Immunogen: Peptide corresponding to the hinge region of human IgG4 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgG4; (-) Human IgG1, IgG2, IgG3, IgM, IgA, IgD, IgE; (-) Cynomolgus monkey, goat, mouse, rat, or rhesus monkey IgG • Applications: ELISA, FC, ICC, IHC, WB • MW = 150 and 50 kDa for whole IgG4 and the IgG4 heavy chain under non-reducing and reducing conditions, respectively