Learn More
Cayman Chemical IgGmouse Monclonal ntIbo 100ug
SDP
Catalog No. 502985934
Encompass_Preferred
Immunogen: Mouse IgG • Host: Goat • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Mouse IgG1, IgG2a, IgG2b, IgG3 (-) Mouse IgM, IgA, IgE (-) Human, rabbit, or rat IgG • Applications: ELISA, FC, IP • MW = None listed
Catalog No. 50-298-5934 Supplier Cayman Chemical Supplier No. 22469100 UG
Immunogen: Mouse IgG • Host: Goat • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Mouse IgG1, IgG2a, IgG2b, IgG3 (-) Mouse IgM, IgA, IgE (-) Human, rabbit, or rat IgG • Applications: ELISA, FC, IP • MW = None listed