Learn More
Cayman Chemical IgGmouse RabbIt MoncLNal 100ug
SDP
Catalog No. 502949754
Encompass_Preferred
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Goat IgG; (+) Mouse IgG1, IgG2a, IgG2b, IgG3; (-) Human, rat IgG; (-) Mouse IgA, IgE, IgM • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-9754 Supplier Cayman Chemical Supplier No. 32002100 UG
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Goat IgG; (+) Mouse IgG1, IgG2a, IgG2b, IgG3; (-) Human, rat IgG; (-) Mouse IgA, IgE, IgM • Applications: ELISA, WB (non-reducing conditions)