Learn More
Cayman Chemical IgGmouse RabbIt Monclonal 50ug
SDP
Catalog No. 502947195
Encompass_Preferred
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: Mouse • Cross Reactivity: (+) IgG1, IgG2a, IgG2b, IgG3 (+) Goat IgG; (-) Mouse IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-7195 Supplier Cayman Chemical Supplier No. 3234850 UG
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: Mouse • Cross Reactivity: (+) IgG1, IgG2a, IgG2b, IgG3 (+) Goat IgG; (-) Mouse IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions)