Learn More
Bioworld IGGMOUSE) SEPAROPOREAGA
Catalog No. 50197871
Encompass
IGG (MOUSE) SEPAROPORE (AGAROSE) 4B: IgG (Mouse) Separopore An affinity chromatography gel matrix consiting of mouse IgG bound to 4% agarose beads. This matrix is coupled through a stable, leak resistant linkage over a wide pH range....
Catalog No. 50-197-871 Supplier Bioworld Supplier No. 201810782
IGG (MOUSE) SEPAROPORE (AGAROSE) 4B: IgG (Mouse) Separopore An affinity chromatography gel matrix consiting of mouse IgG bound to 4% agarose beads. This matrix is coupled through a stable, leak resistant linkage over a wide pH range....