Learn More
Bioworld IGGPURE PROTEIN-A SEPAROPO
Catalog No. 50197920
Encompass
IGGPURE, PROTEIN-A SEPAROPORE (AGAROSE) KIT: IgGPure Protein A Separopore Kit consists ofaffinity matrix with immobilized Protein-A (from Staphylococcus aureus) for purification and/or fractionation of IgG subclasses. Reliable tool to pu...
Catalog No. 50-197-920 Supplier Bioworld Supplier No. 201811011
IGGPURE, PROTEIN-A SEPAROPORE (AGAROSE) KIT: IgGPure Protein A Separopore Kit consists ofaffinity matrix with immobilized Protein-A (from Staphylococcus aureus) for purification and/or fractionation of IgG subclasses. Reliable tool to pu...