Learn More
Cayman Chemical IgGrabbIt Monclonal ntIb 100ug
SDP
Catalog No. 502948190
Encompass_Preferred
Immunogen: Rabbit IgG • Host: Goat • Species Reactivity: (+) Rabbit • Cross Reactivity: (-) Human, mouse, and rat IgG • Applications: ELISA, WB
Catalog No. 50-294-8190 Supplier Cayman Chemical Supplier No. 32105100 UG
Immunogen: Rabbit IgG • Host: Goat • Species Reactivity: (+) Rabbit • Cross Reactivity: (-) Human, mouse, and rat IgG • Applications: ELISA, WB