Learn More
Bioworld IGGRABBIT) SEPAROPOREAG
Supplier:  Bioworld 201810151
Encompass
IGG (RABBIT) SEPAROPORE (AGAROSE) 4B: IgG-Separopore An affinity chromatography gel matrix consiting of rabbit IgG bound to 4% agarose beads. This matrix is coupled through a stable, leak resistant linkage over a wide pH range. This al...
Catalog No. 50-197-695