Learn More
GENOVIS INC IgGZERO, 1000 units
Catalog No. NC0820176
Encompass
IgGZERO (EndoS) is an IgG-specific endoglycosidase acting on complex type N-glycans at the Fc-glycosylation site of IgG. The enzyme acts on the chitobiose core and leaves the core GlcNAc intact.
This item is not returnable. View return policy
Catalog No. NC0820176 Supplier GENOVIS INC Supplier No. A0IZ1010
IgGZERO (EndoS) is an IgG-specific endoglycosidase acting on complex type N-glycans at the Fc-glycosylation site of IgG. The enzyme acts on the chitobiose core and leaves the core GlcNAc intact.