Learn More
Description
Hybridization signals of digoxigenin-labeled polynucleotides appear dark green distinct dot-shaped (distal to the IGH breakpoint region), and dinitrophenyl-labeled polynucleotides appear bright red distinct dot-shaped (proximal to the IGH breakpoint region).
Normal situation: In interphases of normal cells or cells without a translocation involving the IGH locus, two red/green fusion signals appear.
Aberrant situation: One IGH locus affected by a translocation is indicated by one separate green signal and one separate red signal. Genomic aberrations due to small deletions, duplications or inversions might result in inconspicuous signal patterns. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Specifications
| Content And Storage | The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. |
| For Use With (Application) | CISH-Ce |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.