Learn More
Cayman Chemical IgK Light Chain mouse Rabbi
SDP
Catalog No. 502949752
Encompass_Preferred
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Igκ light chain; (-) Mouse Igλ light chain (-) Human, goat, rat IgG; • Applications: ELISA, WB • MW = ~150 and ~25 kDa for non-reduced and reduced Igκ, respectively
Catalog No. 50-294-9752 Supplier Cayman Chemical Supplier No. 3234750 UG
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Igκ light chain; (-) Mouse Igλ light chain (-) Human, goat, rat IgG; • Applications: ELISA, WB • MW = ~150 and ~25 kDa for non-reduced and reduced Igκ, respectively