Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3514MSM8P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: KLC1278. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: 293T, Raji or hPBL cells. Tonsil or Spleen. Cellular Staining: Cell Surface, cytoplasmic and secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length Ig kappa chain. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 22.5kDa.Chromosome: 2p11.2. Gene Symbol: IGKC. Gene ID: 3514.
Catalog No. 50-188-4813