Learn More
Abnova™ IGL Split FISH Probe
Catalog No. 89337322 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
FISH-Ce
Catalog No. For Use With (Application)
89-337-322 FISH-Ce
1 options
Catalog No. 89-337-322 Supplier Abnova™ Supplier No. FS0086
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
For Use With (Application) FISH-Ce