Learn More
Diagnostic Biosystems IGM IgM
Catalog No. 502407575
Encompass_Preferred
This antibody reacts with the heavy chain present in all types of human IgM. It stains B cell follicles in human lymphoid tissues. In lymphoid tissues, IgM containing plasma cells stain strongly but immune complexes are usually undetectable.
Catalog No. 50-240-7575 Supplier Diagnostic Biosystems Supplier No. PDR019
This antibody reacts with the heavy chain present in all types of human IgM. It stains B cell follicles in human lymphoid tissues. In lymphoid tissues, IgM containing plasma cells stain strongly but immune complexes are usually undetectable.