Learn More
Alpco Ltd IgM ELISA 96 Wells 5 L Research Use Only. Not for Use in Diagnostic Procedures.
SDP
Supplier:  Alpco Ltd 41IGMHUE01
Encompass
The IgM ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgM in human biological samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3597