Learn More
Diagnostic Biosystems IGM-FITC IgM-FITC
Supplier:  Diagnostic Biosystems F009
Encompass_Preferred
This antibody reacts with the Mu chain of human IgM. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7463