Learn More
Diagnostic Biosystems IGM-FITC IgM-FITC
Catalog No. 502407527
Encompass_Preferred
This antibody reacts with the Mu chain of human IgM. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7527 Supplier Diagnostic Biosystems Supplier No. F00901
This antibody reacts with the Mu chain of human IgM. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.