Learn More
Sigma Aldrich Fine Chemicals Biosciences IgM from bovine serum reag10MG
Catalog No. 502904549
Encompass
IgM is a highly conserved antibody in vertebrates and is expressed early during immune response. It exists as a pentamer and is secreted by peritoneal B cells.
Catalog No. 50-290-4549 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I813510MG
IgM is a highly conserved antibody in vertebrates and is expressed early during immune response. It exists as a pentamer and is secreted by peritoneal B cells.