Learn More
Sigma Aldrich Fine Chemicals Biosciences IgM from human serum reage5MG
Catalog No. 502914388
Encompass
There are five immunoglobulin classes in humans. Out of these immunoglobulin M (IgM) is a high molecular weight protein that has five or six subunits. IgM monomers are made of two heavy chains and two light chains connected by a disulfide bond. Human serum shows low concentration of IgM monomers. It has a high carbohydrate content of about 12%. It is the first immunoglobulin produced by neonates.IgM antibodies are present as pentamers in the serum and are produced in response to antigens.IgM from human serum is purified from normal human serum by precipitation and gel filtration techniques. The immunoglobulin is determined to be atleast 95% pure by HPLC procedures.
Catalog No. 50-291-4388 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I82605MG
There are five immunoglobulin classes in humans. Out of these immunoglobulin M (IgM) is a high molecular weight protein that has five or six subunits. IgM monomers are made of two heavy chains and two light chains connected by a disulfide bond. Human serum shows low concentration of IgM monomers. It has a high carbohydrate content of about 12%. It is the first immunoglobulin produced by neonates.IgM antibodies are present as pentamers in the serum and are produced in response to antigens.IgM from human serum is purified from normal human serum by precipitation and gel filtration techniques. The immunoglobulin is determined to be atleast 95% pure by HPLC procedures.