Learn More
IgM (H+L) Mouse anti-Rabbit, DyLight 550, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | NRBM |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Rabbit IgM |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.