Learn More
Athens Research And Technology Inc IGM HUMAN PLASMA 1MG
Supplier:  Athens Research And Technology Inc 16160907131MG
Encompass_Preferred
Immunoglobulin M, Human Normal Plasma: 1 MG; Frozen in 50 mM Tris-HCl, pH 8.0, with 200 mM NaCl and 0.05% NaN3; MW 950,000; EC= 1.18; Purity: > =95% by SDS-PAGE. Native; Prepared and purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-820