Learn More
Enzo Life Sciences IgM (mouse), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900120
Encompass_Preferred
The IgM (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Absorbance is read at 450 nm. Ideal for determining the isotype and concentration of monoclonal antibodies. Sensitivity: 0.60 ng/ml (range 3.91 - 250 ng/ml). Assay Time: 90 minutes. Applications: ELISA, Colorimetric detection. Species Reactivity: Mouse. Contents: GxM IgG Microtiter plate, mouse IgM conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgGM standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-155