Learn More
CAPRICO BIOTECHNOLOGIES INC Caprico IGM PE 4113025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4113025
Encompass_Preferred
The Caprico IGM PE 4113025 is a polyclonal antibody produced in goat that is immunoreactive against mouse IgM. This antibody can be used in ELISA, Western Blot, Immunohistochemistry, and Flow Cytometry applications. The immunogen is purified mouse IgM whole molecule. This antibody is supplied as a lyophilized powder with 0.05% sodium azide as a preservative. Each vial contains 100 ug of antibody that should be reconstituted in 1 mL of PBS. The reconstituted antibody can be stored at 4°C for short term storage or aliquoted and stored at -20°C for long term storage. This antibody is optimized for use in mouse samples.
Catalog No. 50-227-9306