Learn More
Innovative Research Inc Human IgM Purified Lyophilized 1mg
Supplier:  Innovative Research Inc IHUIGMLY1MG
Encompass_Preferred
Human IgM Purified Lyophilized from Innovative Research has been purified from human plasma. This is a lyophilized powder buffered in 50 mM tris, 200 mM sodium chloride, and 0.05% sodium azide, pH 8.0, with a purity of greater than 96% as determined by SDS-PAGE analysis.
Catalog No. 50-203-6437