Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Rat anti-Mouse, AF 555, Clone: 1B4B1, Southern Biotech™
SDP

Catalog No. OB114032 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB114032 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB114032 Supplier Southern Biotechnology Associates Supplier No. 114032
Add to Cart
Add to Cart

Rat Monoclonal Secondary Antibody

SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen IgM
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 1B4B1
Concentration 0.5mg/mL
Conjugate Alexa Fluor 555
Formulation Supplied in PBS/NaN3
Host Species Rat
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.