Learn More
INTACT GENOMICS INC igMax™ 10B ElectroCompetent Cells 6x100l
Catalog No. 5000082953
Encompass
Intact Genomics igMax™ 10B derivative electrocompetent cells are suitable for demanding cloning situations such as synthetic bio-applications, BAC cloning, assembling large multi-DNA fragments, or cloning difficult targets requiring the greatest number of transformants possible. Utilizing proprietary manufacturing methods, these cells allow for effective transformation of all large DNA molecules (10kb up to 350kb)!Product Include and Storage:igMax™ 10B Electrocompetent cells: -80 CpUC19 control DNA: -20 CRecovery medium: 4 C
Catalog No. 50-000-82953 Supplier INTACT GENOMICS INC Supplier No. 128424
Intact Genomics igMax™ 10B derivative electrocompetent cells are suitable for demanding cloning situations such as synthetic bio-applications, BAC cloning, assembling large multi-DNA fragments, or cloning difficult targets requiring the greatest number of transformants possible. Utilizing proprietary manufacturing methods, these cells allow for effective transformation of all large DNA molecules (10kb up to 350kb)!Product Include and Storage:igMax™ 10B Electrocompetent cells: -80 CpUC19 control DNA: -20 CRecovery medium: 4 C