Learn More
Medchemexpress LLC (+)-Igmesine hydrochloride | 00-00-0 | 99.8% | 355.94 g·mol⁻¹ | C23H30ClN | 5 MG
SDP
Catalog No. 5000264136
Encompass_Preferred
(+)-Igmesine hydrochloride is the hydrochloride salt of igmesine, a selective sigma receptor ligand reported to have neuroprotective and anti-amnesic activity in preclinical studies. It is provided for research use only and is characterized by a defined molecular formula (C23H30ClN) and molecular weight (355.94 g·mol⁻¹).
  • Selective sigma receptor ligand.
  • Demonstrated neuroprotective and anti-amnesic activity in models.
  • Hydrochloride salt for improved solubility and handling.
  • High purity (99.8%).
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store protected from light and under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-64136 Supplier Medchemexpress LLC Supplier No. HY1085155MG
(+)-Igmesine hydrochloride is the hydrochloride salt of igmesine, a selective sigma receptor ligand reported to have neuroprotective and anti-amnesic activity in preclinical studies. It is provided for research use only and is characterized by a defined molecular formula (C23H30ClN) and molecular weight (355.94 g·mol⁻¹).
  • Selective sigma receptor ligand.
  • Demonstrated neuroprotective and anti-amnesic activity in models.
  • Hydrochloride salt for improved solubility and handling.
  • High purity (99.8%).
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store protected from light and under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).