Learn More
Medchemexpress LLC (+)-Igmesine hydrochloride | 130152-35-1 | 355.9 | 50 MG
SDP
Catalog No. 5000345237
Encompass_Preferred
(+)-Igmesine hydrochloride is an orally active and selective sigma receptor ligand with neuroprotective and antidepressant properties.
Catalog No. 50-003-45237 Supplier Medchemexpress LLC Supplier No. HY1085151MG
(+)-Igmesine hydrochloride is an orally active and selective sigma receptor ligand with neuroprotective and antidepressant properties.