Learn More
Cayman Chemical IgMhumn RabbIt Monclonal 50ug
SDP
Catalog No. 502946583
Encompass_Preferred
Immunogen: Human IgM • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgM; (-) Human IgG, IgA, IgD, or IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6583 Supplier Cayman Chemical Supplier No. 3236750 UG
Immunogen: Human IgM • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgM; (-) Human IgG, IgA, IgD, or IgE • Applications: ELISA, ICC, IHC