Learn More
Cayman Chemical Igmouse RabbIt Monclonal 100ug
SDP
Catalog No. 502948082
Encompass_Preferred
Immunogen: Mouse IgM • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE (-) Human, rat, goat IgG • Applications: ELISA, FC, IP
Catalog No. 50-294-8082 Supplier Cayman Chemical Supplier No. 32101100 UG
Immunogen: Mouse IgM • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE (-) Human, rat, goat IgG • Applications: ELISA, FC, IP