Learn More
Cayman Chemical Igmouse RabbIt Monclonal 50ug
SDP
Catalog No. 502950083
Encompass_Preferred
Immunogen: Mouse IgM • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgM; (-) Mouse IgG1, IgG2a, IgG2b, IgA, IgE; (-) Human, goat, rat IgG • Applications: ELISA
Catalog No. 50-295-0083 Supplier Cayman Chemical Supplier No. 3235650 UG
Immunogen: Mouse IgM • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgM; (-) Mouse IgG1, IgG2a, IgG2b, IgA, IgE; (-) Human, goat, rat IgG • Applications: ELISA