Learn More
INTACT GENOMICS INC igScript™ Probe-Based qPCR master mix 500 reactions
Supplier:  INTACT GENOMICS INC 4233
Encompass
IgScript™ Probe-Based qPCR 2x master mix contains IgScript™ Taq DNA polymerase, MgCl2, dNTPs, stabilizers, enhancers and low ROX reference dye in a standard buffer. It provides improved qPCR efficiency, wider dynamic range, superior sensitivity and specificity. IgScript™ Probe-Based qPCR 2x master mix is a ready-to-use cocktail containing all components except primers, probe and template, for the amplification and detection of DNA in qPCR. This 2x master mix requires minimal handling during reaction setup. Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase (1, 2) and a 5´→3´ exonuclease activity (3, 4). The amplification step features a high quality Taq DNA Polymerase Enhanced efficiency, specificity, and sensitivity. Compatible with all real-time PCR instruments for Gene expression validation, multiplexing, mutation detection, pathogen detection and GMO characterization.
Catalog No. 50-000-82845