Learn More
Aviva Systems Biology Corp IGSF1 Antibody - middle region (ARP42748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGSF1. Alternate names: IGCD1, IGDC1, INHBP, KIAA0364, MGC75490, PGSF2, p120ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58326