Learn More
Aviva Systems Biology Corp IGSF11 Antibody - middle region (ARP44704_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44704_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGSF11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGSF11. Alternate names: BT-IgSF, CXADRL1, Igsf13, MGC35227, VSIG3, CT119ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58878