Learn More
Aviva Systems Biology Corp IGSF8 Antibody - middle region (ARP59875_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGSF8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human IGSF8. Alternate names: CD316, CD81P3, EWI2, PGRL, EWI-2, KCT-4, LIR-D1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit
Catalog No. 50-286-862