Learn More
Alpco Ltd Chicken IgY ELISA
SDP
Supplier:  Alpco Ltd 41IGYCHE01
Encompass
The Chicken IgY ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgY in serum or plasma of chickens. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3600