Learn More
Newcomer Supply TOMO Adhesive Microscope Slides, Pink, 1000/case
SDP
Catalog No. NC1011921
Encompass
The advanced technology in manufacturing the adhesive surface of the TOMO microscope slide produces outstanding adhesion to the tissue section even in the most demanding applications. The surface of the TOMO adhesive microscope slide is hydrophilic producing two very notable features: allows to refloat the tissue in the water bath for repositioning the tissue section and allows the microscope slide to be used on all automated IHC platforms. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 45° Corner (90° Corners available in White); Available Colors: White, Blue, Green, Yellow and Pink; Unit: 100 slides/box, 1,000 slides/case. ISO 13485 Certified.
Catalog No. NC1011921 Supplier Newcomer Supply Supplier No. 5081P
The advanced technology in manufacturing the adhesive surface of the TOMO microscope slide produces outstanding adhesion to the tissue section even in the most demanding applications. The surface of the TOMO adhesive microscope slide is hydrophilic producing two very notable features: allows to refloat the tissue in the water bath for repositioning the tissue section and allows the microscope slide to be used on all automated IHC platforms. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 45° Corner (90° Corners available in White); Available Colors: White, Blue, Green, Yellow and Pink; Unit: 100 slides/box, 1,000 slides/case. ISO 13485 Certified.