Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ IHC Antigen Retrieval Solution - Low pH (10X)

Catalog No. 5018782
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-187-82 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-187-82 Supplier Invitrogen™ Supplier No. 00495558
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin-embedded tissue sections. Used in combination with heat has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue.

  • Supplied as a 10x stock solution
  • Dilution of this buffer to 1x using reagent-grade water results in a 10 mM Sodium Citrate solution, pH 6.0
TRUSTED_SUSTAINABILITY

Specifications

Product Type IHC Antigen Retrieval Solution
Quantity 500 mL
For Use With (Application) Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.