Learn More
Leica Biosystems IHC Diluent
Supplier:  Leica Biosystems RE7133CE
Encompass
Novocastra IHC Diluent is intended for use as a diluent for Novocastra primary antibodies in immunohistochemical (IHC) procedures. Novocastra IHC Diluent is not intended for the reconstitution of lyophilized antibodies. Volume supplied is 500 mL. For in vitro diagnostic use.
Catalog No. 50-255-1987