Learn More
Enzo Life Sciences IHC PAP pen (1 ea)
SDP
Catalog No. NC1384848
Encompass_Preferred
The IHC PAP pen provides a hydrophobic barrier when a circle is drawn around a specimen on a glass slide. Applications: IF, IHC, ISH (in situ hybridization). Shipping: Ambient. Long Term Storage: Ambient
Catalog No. NC1384848 Supplier Enzo Life Sciences Supplier No. ADI9502330001
The IHC PAP pen provides a hydrophobic barrier when a circle is drawn around a specimen on a glass slide. Applications: IF, IHC, ISH (in situ hybridization). Shipping: Ambient. Long Term Storage: Ambient