Learn More
Enzo Life Sciences IHC PBS tablets (100ea)
SDP
Supplier:  Enzo Life Sciences ADI9502400100
Encompass_Preferred
Formulation: Solid. 100 ml PBS (1X) tablets containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer. Long Term Storage: Ambient. Handling: Protect from moisture. Applications: IHC .
Catalog No. 50-200-7119