Learn More
Vector Laboratories IHC‑plus™ Monoclonal Mouse anti‑Human IL2RA / CD25 Antibody (IHC, WB) LS‑B7396, 500ug
SDP
Supplier:  Vector Laboratories LSB7396500
Encompass
CD25 antibody LS-B7396 is an unconjugated mouse monoclonal antibody to human CD25 (IL2RA). Validated for ELISA, IHC and WB. Tested on 20 paraffin-embedded human tissues.
Catalog No. NC2681013