Learn More
Enzo Life Sciences POLYVIEW IHC reagent (mouse/rabbit-HRP) (100tests)
SDP
Supplier:  Enzo Life Sciences ADI9501130100
Encompass_Preferred
High sensitivity, low background nanopolymer detection reagent for use with HIGHDEF chromogens. The POLYVIEW (mouse/rabbit-HRP) IHC detection reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The HRP detection reagent may also be used with blood smears, cytosmears, and cell preparations. Content: Anti-Mouse/Rabbit HRP detection reagent. Long Term Storage: +4°C. Applications: IHC .
Catalog No. 50-200-7115