Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IHC Select™ Secondary Goat Rabbit IgG Goat anti-Rabbit, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 21537MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-537-MI 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-537-MI Supplier MilliporeSigma Supplier No. 21537
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat IgG antibody is validated for use in immunohistochemistry for the detection of the Rabbit IgG protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation Goat anti-Rabbit IgG in PBS containing carrier protein, with 0.1% ProClin 300 as preservative.
Host Species Goat
Quantity 15 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C in undiluted format. Do not Freeze.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.