Learn More
MilliporeSigma IHC Select HRP/DAB, 150 Test
Catalog No. NC9574428
Encompass
IHC Select HRP/DAB Tests for Immunocytochemistry, Immunohistochemistry & Immunohistochemistry (Paraffin). The IHC Select Immunoperoxidase Secondary Detection System is intended for use with the CHEMICON IHC Select Prediluted Primary Antibody reagents which contain rabbit and mouse IgG for the qualitative identification of antigens by light microscopy in paraffin-embedded tissues. The IHC Select Immunoperoxidase Secondary Detection System may also be used with other rabbit or mouse IgG primary antibodies.
Catalog No. NC9574428 Supplier MilliporeSigma Supplier No. DAB150
IHC Select HRP/DAB Tests for Immunocytochemistry, Immunohistochemistry & Immunohistochemistry (Paraffin). The IHC Select Immunoperoxidase Secondary Detection System is intended for use with the CHEMICON IHC Select Prediluted Primary Antibody reagents which contain rabbit and mouse IgG for the qualitative identification of antigens by light microscopy in paraffin-embedded tissues. The IHC Select Immunoperoxidase Secondary Detection System may also be used with other rabbit or mouse IgG primary antibodies.