Learn More
Leica Biosystems IHC SUPER BLOCKING 110ML
Catalog No. NC0336255
Encompass_Preferred
IHC/ISH Super Blocking is specially formulated to stabilize antibodies and eliminate background encountered in immunohistochemistry (IHC) and in situ hybridization (ISH) procedures. The diluent contains complex protein and non-protein mixtures that eliminate background by blocking non-specific protein-protein interactions. It is particularly recommended for staining of mouse tissues with mouse primary antibodies. It can also be used as a diluent for primary antibodies. If primary antibodies are diluted in this solution, a blocking step before primary antibodies can be omitted.
Catalog No. NC0336255 Supplier Leica Biosystems Supplier No. PV6122
IHC/ISH Super Blocking is specially formulated to stabilize antibodies and eliminate background encountered in immunohistochemistry (IHC) and in situ hybridization (ISH) procedures. The diluent contains complex protein and non-protein mixtures that eliminate background by blocking non-specific protein-protein interactions. It is particularly recommended for staining of mouse tissues with mouse primary antibodies. It can also be used as a diluent for primary antibodies. If primary antibodies are diluted in this solution, a blocking step before primary antibodies can be omitted.