Learn More
Ihc World LLC IHC-TEK EPITOPE SOLUTION 1L
SDP
Catalog No. NC0773482
Encompass
This Newly Formulated Solution Is To Be Used Universally For Retrieving Antigens And Epitopes Of All Types, Including Membrane Proteins, Cytoplasmic Proteins As Well As Nuclear Proteins, On Formalin Fixed Paraffin Embedded Tissue Sections During Immunohistochemistry Procedure. This Solution Is Superior To Commonly Used Citrate Based Solutions And Less Tissue Destruction Than Other High Ph Solutions.
Catalog No. NC0773482 Supplier Ihc World LLC Supplier No. IW11001L
This Newly Formulated Solution Is To Be Used Universally For Retrieving Antigens And Epitopes Of All Types, Including Membrane Proteins, Cytoplasmic Proteins As Well As Nuclear Proteins, On Formalin Fixed Paraffin Embedded Tissue Sections During Immunohistochemistry Procedure. This Solution Is Superior To Commonly Used Citrate Based Solutions And Less Tissue Destruction Than Other High Ph Solutions.