Learn More
Ihc World LLC IHC TEK PROTEINASE K 20ML
SDP
Catalog No. NC0056220
Encompass
Formalin or other aldehyde fixation forms protein cross-links that mask the antigenic sites in tissue specimens, thereby giving weak or false negative staining for immunohistochemical detection of certain proteins. This proteinase K based solution ca
Catalog No. NC0056220 Supplier Ihc World LLC Supplier No. IW1101
Formalin or other aldehyde fixation forms protein cross-links that mask the antigenic sites in tissue specimens, thereby giving weak or false negative staining for immunohistochemical detection of certain proteins. This proteinase K based solution ca