Learn More
Ihc World LLC IHC-TEKTM ANTIBODY DILUENT PH
SDP
Catalog No. NC9953741
Encompass_Preferred
This PBS based antibody diluent can be used to dilute both primary and secondary antibodies for IHC, ICC, IF and whole mount staining. Serum blocking step can be omitted when using this product and it is particularly useful for enhancing staining int
Catalog No. NC9953741 Supplier Ihc World LLC Supplier No. IW1000
This PBS based antibody diluent can be used to dilute both primary and secondary antibodies for IHC, ICC, IF and whole mount staining. Serum blocking step can be omitted when using this product and it is particularly useful for enhancing staining int